siRNA Details
| Gene Symbol: CCDC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135197.1 | NP_001128669.1 | 8 | 1171 | ||
| NM_178173.3 | NP_835467.2 | 10 | 1469 | |||
| XM_006713124.3 | XP_006713187.1 | 8 | 1346 | |||
| XM_011533670.2 | XP_011531972.1 | 7 | 1180 | |||
| XM_011533671.2 | XP_011531973.1 | 7 | 1174 | |||
| XM_011533672.2 | XP_011531974.1 | 8 | 1539 | |||
| XM_011533673.2 | XP_011531975.1 | 8 | 1571 | |||
| GenBank mRNA | AK058049.1 | - | 10 | 1360 | ||
| AK303033.1 | - | 8 | 1171 | |||
| BC036202.1 | - | 10 | 1469 | 4836976 | ||

CCDC36
Genomic Map