siRNA Details
| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182702.1 | NP_874361.1 | 3 | 471 | ||
| GenBank mRNA | AJ544583.1 | - | 3 | 471 | ||
| AK092703.1 | - | 2 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS42| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182702.1 | NP_874361.1 | 3 | 471 | ||
| GenBank mRNA | AJ544583.1 | - | 3 | 471 | ||
| AK092703.1 | - | 2 | 215 | |||