siRNA Details
| Gene Symbol: TMPRSS11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114387.1 | NP_001107859.1 | 7 | 656 | ||
| NM_182606.3 | NP_872412.3 | 7 | 665 | |||
| GenBank mRNA | AK131518.1 | - | 7 | 665 | ||
| AK292950.1 | - | 7 | 594 | |||
| AY498712.1 | - | 7 | 568 | |||
| BC111796.1 | - | 7 | 582 | 40026103 | ||

TMPRSS11A
Genomic Map