siRNA Details
| Gene Symbol: SLC6A19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003841.2 | NP_001003841.1 | 3 | 437 | ||
| GenBank mRNA | AK290811.1 | - | 3 | 437 | ||
| AY591756.1 | - | 3 | 381 | |||
| AY596807.1 | - | 3 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC6A19| Gene Symbol: SLC6A19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003841.2 | NP_001003841.1 | 3 | 437 | ||
| GenBank mRNA | AK290811.1 | - | 3 | 437 | ||
| AY591756.1 | - | 3 | 381 | |||
| AY596807.1 | - | 3 | 381 | |||