siRNA Details
| Gene Symbol: FAM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163991.1 | NP_001157463.1 | 1 | 266 | ||
| NM_182761.3 | NP_877438.2 | 1 | 266 | |||
| GenBank mRNA | AF427126.1 | - | 1 | 266 | ||
| AY692447.1 | - | 1 | 266 | |||
| BC040622.1 | - | 1 | 227 | |||
| BC128242.1 | - | 1 | 139 | |||
| BC128243.1 | - | 1 | 139 | 40114744 | ||

FAM170A
Genomic Map