siRNA Details
| Gene Symbol: ARSI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012301.2 | NP_001012301.1 | 1 | 784 | ||
| GenBank mRNA | AB448735.1 | - | 1 | 775 | ||
| AY875937.1 | - | 1 | 195 | |||
| BC129995.1 | - | 1 | 252 | 40127932 | ||
| BC129996.1 | - | 1 | 252 | 40127934 | ||

ARSI
Genomic Map