siRNA Details
| Gene Symbol: MYLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012418.3 | NP_001012418.2 | 7 | 877 | ||
| XM_005249078.3 | XP_005249135.2 | 7 | 842 | |||
| XM_005249079.2 | XP_005249136.1 | 7 | 735 | |||
| XM_006715082.3 | XP_006715145.1 | 6 | 2172 | |||
| XM_011514567.2 | XP_011512869.1 | 7 | 965 | |||
| XM_017010819.1 | XP_016866308.1 | 7 | 774 | |||
| GenBank mRNA | BC132831.1 | - | 6 | 643 | 40146853 | |
| BC132833.1 | - | 6 | 643 | 40146855 | ||
| BX647582.1 | - | 7 | 877 | |||

MYLK4
Genomic Map