siRNA Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271807.1 | NP_001258736.1 | 2 | 417 | ||
| NM_178174.3 | NP_835468.1 | 2 | 417 | |||
| XM_017010822.1 | XP_016866311.1 | 4 | 607 | |||
| XM_017010823.1 | XP_016866312.1 | 3 | 495 | |||
| XM_017010824.1 | XP_016866313.1 | 3 | 501 | |||
| XM_017010825.1 | XP_016866314.1 | 4 | 607 | |||
| GenBank mRNA | AF508193.1 | - | 2 | 356 | ||
| AF534822.1 | - | 2 | 400 | |||
| AF534823.1 | - | 2 | 400 | |||
| AY358357.1 | - | 2 | 405 | |||
| BC100944.1 | - | 2 | 400 | 40004190 | ||
| BC100945.1 | - | 2 | 400 | 40004191 | ||

TREML1
Genomic Map