siRNA Details
| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178562.4 | NP_848657.1 | 8 | 1050 | ||
| XM_006715960.3 | XP_006716023.1 | 8 | 878 | |||
| GenBank mRNA | AF276891.1 | - | 8 | 785 | ||
| AK000208.1 | - | 3 | 664 | |||
| AY236849.1 | - | 8 | 874 | |||
| BC044244.1 | - | 8 | 1014 | 5759132 | ||

TSPAN33
Genomic Map