siRNA Details
| Gene Symbol: NHSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013627.2 | NP_001013649.2 | 6 | 1729 | ||
| XM_011530932.1 | XP_011529234.1 | 7 | 1813 | |||
| XM_011530933.1 | XP_011529235.1 | 6 | 1681 | |||
| XM_011530934.2 | XP_011529236.1 | 6 | 6521 | |||
| XM_017029473.1 | XP_016884962.1 | 6 | 5491 | |||
| XM_017029474.1 | XP_016884963.1 | 6 | 1633 | |||
| XM_017029475.1 | XP_016884964.1 | 2 | 4876 | |||
| XM_017029476.1 | XP_016884965.1 | 2 | 993 | |||
| XM_017029477.1 | XP_016884966.1 | 2 | 5907 | |||
| GenBank mRNA | BC033261.1 | - | 2 | 1893 | ||
| BC136756.1 | - | 3 | 1102 | 9020746 | ||

NHSL2
Genomic Map