siRNA Details
| Gene Symbol: ZCCHC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004308.2 | NP_001004308.2 | 3 | 598 | ||
| GenBank mRNA | AK128465.1 | - | 3 | 598 | ||
| BC137430.1 | - | 1 | 391 | 9021428 | ||
| BC137433.1 | - | 1 | 391 | 9021431 | ||

ZCCHC16
Genomic Map