siRNA Details
| Gene Symbol: MS4A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206893.3 | NP_996776.2 | 6, 7 | 707 | ||
| XM_011544989.1 | XP_011543291.1 | 6, 7 | 707 | |||
| XM_011544991.2 | XP_011543293.1 | 5, 6 | 518 | |||
| GenBank mRNA | AK122633.1 | - | 6, 7 | 708 | ||
| BC137259.1 | - | 6, 7 | 657 | 9021256 | ||
| BC137263.1 | - | 6, 7 | 657 | 9021260 | ||

MS4A10
Genomic Map