siRNA Details
| Gene Symbol: SMCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145010.1 | NP_001138482.1 | 6, 7 | 784 | ||
| XM_011520634.2 | XP_011518936.1 | 6, 7 | 962 | |||
| XM_011520635.2 | XP_011518937.1 | 4, 5 | 461 | |||
| XM_017019254.1 | XP_016874743.1 | 4, 5 | 718 | |||
| XM_017019255.1 | XP_016874744.1 | 3, 4 | 568 | |||

SMCO2
Genomic Map