siRNA Details
| Gene Symbol: SMCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145010.1 | NP_001138482.1 | 5 | 558 | ||
| XM_011520634.2 | XP_011518936.1 | 5 | 736 | |||
| XM_011520635.2 | XP_011518937.1 | 3 | 235 | |||
| XM_011520636.2 | XP_011518938.1 | 5 | 736 | |||
| XM_017019254.1 | XP_016874743.1 | 3 | 492 | |||

SMCO2
Genomic Map