siRNA Details
| Gene Symbol: OVCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183378.2 | NP_899234.2 | 9 | 1038 | ||
| XM_011520638.2 | XP_011518940.1 | 10 | 3189 | |||
| XM_011520639.2 | XP_011518941.1 | 10 | 3189 | |||
| XM_011520640.2 | XP_011518942.1 | 10 | 3189 | |||
| XM_011520641.2 | XP_011518943.1 | 10 | 3189 | |||
| XM_011520642.2 | XP_011518944.1 | 10 | 3189 | |||
| XM_017019256.1 | XP_016874745.1 | 10 | 3189 | |||
| XM_017019257.1 | XP_016874746.1 | 10 | 3189 | |||

OVCH1
Genomic Map