siRNA Details
| Gene Symbol: ZNF774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004309.2 | NP_001004309.2 | 3 | 307 | ||
| XM_017022127.1 | XP_016877616.1 | 4 | 315 | |||
| XM_017022128.1 | XP_016877617.1 | 2 | 989 | |||
| XM_017022129.1 | XP_016877618.1 | 3 | 172 | |||
| GenBank mRNA | AK289385.1 | - | 3 | 307 | ||
| BC067279.1 | - | 3 | 242 | 6181677 | ||
| CR749343.1 | - | 3 | 212 | |||
| DA017564.1 | - | 3 | 308 | |||

ZNF774
Genomic Map