siRNA Details
| Gene Symbol: ZNF774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004309.2 | NP_001004309.2 | 4 | 677 | ||
| XM_017022127.1 | XP_016877616.1 | 5 | 685 | |||
| XM_017022128.1 | XP_016877617.1 | 3 | 1359 | |||
| XM_017022129.1 | XP_016877618.1 | 4 | 549 | |||
| GenBank mRNA | AK289385.1 | - | 4 | 677 | ||
| BC067279.1 | - | 4 | 612 | 6181677 | ||

ZNF774
Genomic Map