siRNA Details
Gene Symbol: ZNF774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004309.2 | NP_001004309.2 | 4 | 850 | ||
XM_017022127.1 | XP_016877616.1 | 5 | 858 | |||
XM_017022128.1 | XP_016877617.1 | 3 | 1532 | |||
XM_017022129.1 | XP_016877618.1 | 4 | 722 | |||
GenBank mRNA | AK289385.1 | - | 4 | 850 | ||
BC067279.1 | - | 4 | 785 | 6181677 |
