siRNA Details
| Gene Symbol: C16orf96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145011.1 | NP_001138483.1 | 9 | 2520 | ||
| XM_005255298.3 | XP_005255355.1 | 10 | 3047 | |||
| XM_011522475.2 | XP_011520777.1 | 11 | 3030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf96| Gene Symbol: C16orf96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145011.1 | NP_001138483.1 | 9 | 2520 | ||
| XM_005255298.3 | XP_005255355.1 | 10 | 3047 | |||
| XM_011522475.2 | XP_011520777.1 | 11 | 3030 | |||