siRNA Details
| Gene Symbol: PKD1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181536.1 | NP_853514.1 | 14 | 2252 | ||
| XM_017023201.1 | XP_016878690.1 | 14 | 2675 | |||
| XM_017023202.1 | XP_016878691.1 | 14 | 2675 | |||
| XM_017023203.1 | XP_016878692.1 | 14 | 2675 | |||
| XM_017023204.1 | XP_016878693.1 | 14 | 2675 | |||
| GenBank mRNA | AY164485.1 | - | 14 | 2252 | ||

PKD1L3
Genomic Map