siRNA Details
| Gene Symbol: SLFN12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195790.1 | NP_001182719.1 | 1, 2 | 1085 | ||
| XM_017024005.1 | XP_016879494.1 | 3, 4 | 2051 | |||
| XM_017024006.1 | XP_016879495.1 | 8, 9 | 4253 | |||
| XM_017024007.1 | XP_016879496.1 | 6, 7 | 3654 | |||
| XM_017024008.1 | XP_016879497.1 | 6, 7 | 7836 | |||
| XM_017024009.1 | XP_016879498.1 | 5, 6 | 7766 | |||
| GenBank mRNA | AK172761.1 | - | 3, 4 | 1886 | ||

SLFN12L
Genomic Map