siRNA Details
| Gene Symbol: SLFN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129820.1 | NP_001123292.1 | 3 | 1438 | ||
| XM_017024576.1 | XP_016880065.1 | 5 | 1607 | |||
| XM_017024577.1 | XP_016880066.1 | 5 | 1629 | |||
| XM_017024578.1 | XP_016880067.1 | 4 | 1496 | |||
| XM_017024579.1 | XP_016880068.1 | 4 | 1529 | |||
| GenBank mRNA | BC140847.1 | - | 3 | 1435 | 9021715 | |
| BC157877.1 | - | 3 | 1439 | 9057094 | ||
| BC157879.1 | - | 3 | 1446 | 9057095 | ||

SLFN14
Genomic Map