siRNA Details
| Gene Symbol: ZNF677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317998.1 | NP_001304927.1 | 5 | 1361 | ||
| NM_182609.3 | NP_872415.1 | 5 | 1179 | |||
| XM_005258862.4 | XP_005258919.1 | 5 | 1395 | |||
| XM_011526912.2 | XP_011525214.1 | 5 | 1307 | |||
| GenBank mRNA | AK308120.1 | - | 5 | 1140 | ||
| BC050038.1 | - | 5 | 1134 | 5272569 | ||
| BX648243.1 | - | 5 | 1150 | |||

ZNF677
Genomic Map