siRNA Details
| Gene Symbol: ZSCAN22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321116.1 | NP_001308045.1 | 3 | 1460 | ||
| NM_001321117.1 | NP_001308046.1 | 3 | 1449 | |||
| NM_181846.2 | NP_862829.1 | 3 | 1465 | |||
| XM_006723192.3 | XP_006723255.1 | 3 | 1437 | |||
| XM_011526917.2 | XP_011525219.1 | 3 | 2543 | |||
| XM_011526918.2 | XP_011525220.1 | 2 | 983 | |||
| XM_017026746.1 | XP_016882235.1 | 3 | 1463 | |||
| XM_017026747.1 | XP_016882236.1 | 3 | 1416 | |||
| GenBank mRNA | AK128716.1 | - | 3 | 1455 | ||
| BC101630.1 | - | 2 | 1378 | 8069136 | ||
| BC112277.1 | - | 2 | 1377 | 8327745 | ||
| BC143446.1 | - | 2 | 1362 | |||
| BX537741.1 | - | 3 | 1465 | |||

ZSCAN22
Genomic Map