siRNA Details
| Gene Symbol: PRAMEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013407.2 | NP_001013425.2 | 3 | 921 | ||
| GenBank mRNA | BC101344.1 | - | 3 | 907 | 40025152 | |
| BC130274.1 | - | 3 | 906 | 40146305 | ||
| BC130276.1 | - | 3 | 906 | 40146307 | ||
| Gene Symbol: PRAMEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010889.2 | NP_001010889.1 | 3 | 907 | ||
| GenBank mRNA | BC101342.2 | - | 3 | 907 | 40025149 | |

PRAMEF5
Genomic Map