siRNA Details
| Gene Symbol: OR6F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005286.1 | NP_001005286.1 | 1 | 881 | ||
| GenBank mRNA | BC137142.1 | - | 1 | 929 | 9021139 | |
| BC137143.1 | - | 1 | 929 | 9021140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6F1| Gene Symbol: OR6F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005286.1 | NP_001005286.1 | 1 | 881 | ||
| GenBank mRNA | BC137142.1 | - | 1 | 929 | 9021139 | |
| BC137143.1 | - | 1 | 929 | 9021140 | ||