siRNA Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287819.1 | NP_001274748.1 | 14 | 1491 | ||
| NM_001287820.1 | NP_001274749.1 | 14 | 1491 | |||
| NM_198503.3 | NP_940905.2 | 14 | 1491 | |||
| XM_006711294.2 | XP_006711357.1 | 14 | 1965 | |||
| XM_006711295.2 | XP_006711358.1 | 14 | 1965 | |||
| XM_011509482.2 | XP_011507784.1 | 13 | 1890 | |||
| XM_011509483.2 | XP_011507785.1 | 14 | 1965 | |||
| XM_017001178.1 | XP_016856667.1 | 14 | 1965 | |||
| XM_017001179.1 | XP_016856668.1 | 14 | 1965 | |||
| XM_017001180.1 | XP_016856669.1 | 14 | 1965 | |||
| XM_017001181.1 | XP_016856670.1 | 4 | 480 | |||
| XM_017001185.1 | XP_016856674.1 | 13 | 1890 | |||
| GenBank mRNA | AK127807.1 | - | 7 | 841 | ||
| AK131498.1 | - | 13 | 1545 | |||
| AK309729.1 | - | 14 | 1480 | |||
| AY359444.1 | - | 14 | 1429 | |||
| BC103947.1 | - | 10 | 872 | |||
| BC103948.1 | - | 14 | 1419 | 40009146 | ||
| BC103949.1 | - | 14 | 1419 | 40009147 | ||
| BC103950.1 | - | 14 | 1419 | 40009148 | ||
| BX647852.1 | - | 14 | 1473 | |||
| EU251076.1 | - | 14 | 1369 | |||

KCNT2
Genomic Map