siRNA Details
| Gene Symbol: OR2T29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004694.2 | NP_001004694.2 | 1 | 656 | ||
| Gene Symbol: OR2T5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004697.1 | NP_001004697.1 | 1 | 656 | ||
| GenBank mRNA | BC140747.1 | - | 1 | 661 | 9021615 | |
| BC140915.1 | - | 1 | 681 | 9056727 | ||

OR2T29
Genomic Map