siRNA Details
| Gene Symbol: NOTO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134462.1 | NP_001127934.1 | 2 | 896 | ||
| GenBank mRNA | AK295936.1 | - | 2 | 896 | ||
| AK316410.1 | - | 2 | 852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOTO| Gene Symbol: NOTO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134462.1 | NP_001127934.1 | 2 | 896 | ||
| GenBank mRNA | AK295936.1 | - | 2 | 896 | ||
| AK316410.1 | - | 2 | 852 | |||