siRNA Details
| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001448.2 | NP_001439.2 | 1 | 290 | ||
| GenBank mRNA | AF030186.1 | - | 1 | 290 | ||
| AF064826.1 | - | 1 | 195 | |||
| AK312605.1 | - | 1 | 547 | |||
| AY052833.1 | - | 1 | 214 | |||
| AY358507.1 | - | 1 | 576 | |||
| BC017166.1 | - | 1 | 584 | 3922774 | ||

GPC4
Genomic Map