siRNA Details
| Gene Symbol: CDKL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009565.1 | NP_001009565.1 | 5 | 482 | ||
| XM_011532815.2 | XP_011531117.1 | 6 | 1898 | |||
| XM_011532816.2 | XP_011531118.1 | 6 | 1898 | |||
| XM_017003981.1 | XP_016859470.1 | 6 | 1250 | |||
| XM_017003982.1 | XP_016859471.1 | 6 | 726 | |||
| XM_017003983.1 | XP_016859472.1 | 7 | 2007 | |||
| GenBank mRNA | AY845084.1 | - | 5 | 482 | ||
| AY847283.1 | - | 5 | 482 | |||
| BC144460.1 | - | 6 | 655 | |||

CDKL4
Genomic Map