siRNA Details
| Gene Symbol: SH3RF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099289.2 | NP_001092759.1 | 8 | 2125 | ||
| XM_011511109.2 | XP_011509411.1 | 8 | 2227 | |||
| XM_011511110.2 | XP_011509412.1 | 6 | 1927 | |||
| GenBank mRNA | AK074131.1 | - | 6 | 1200 | ||
| AL831825.1 | - | 1 | 113 | |||

SH3RF3
Genomic Map