siRNA Details
| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001448.2 | NP_001439.2 | 4 | 1072 | ||
| GenBank mRNA | AF030186.1 | - | 4 | 1072 | ||
| AF064826.1 | - | 4 | 977 | |||
| AK304207.1 | - | 4 | 797 | |||
| AK312605.1 | - | 4 | 1329 | |||
| AY052833.1 | - | 4 | 996 | |||
| AY358507.1 | - | 4 | 1358 | |||
| BC017166.1 | - | 4 | 1366 | 3922774 | ||

GPC4
Genomic Map