siRNA Details
| Gene Symbol: GPR148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207364.2 | NP_997247.2 | 1 | 1192 | ||
| GenBank mRNA | AY569570.1 | - | 1 | 1112 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR148| Gene Symbol: GPR148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207364.2 | NP_997247.2 | 1 | 1192 | ||
| GenBank mRNA | AY569570.1 | - | 1 | 1112 | ||