siRNA Details
| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028301.1 | 1 | 133 | |||
| NR_028302.1 | 1 | 133 | ||||
| GenBank mRNA | AK309100.1 | - | 1 | 133 | ||
| BC039550.1 | - | 1 | 100 | |||
| DA339707.1 | - | 1 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUBR| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028301.1 | 1 | 133 | |||
| NR_028302.1 | 1 | 133 | ||||
| GenBank mRNA | AK309100.1 | - | 1 | 133 | ||
| BC039550.1 | - | 1 | 100 | |||
| DA339707.1 | - | 1 | 133 | |||