siRNA Details
| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038705.1 | NP_001033794.1 | 3 | 1692 | ||
| GenBank mRNA | AY255534.1 | - | 3 | 1127 | ||
| DQ318360.1 | - | 3 | 1692 | |||
More Information
Set Membership: |
|

GPR149
Genomic Map