siRNA Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005473.2 | NP_001005473.1 | 3 | 1024 | ||
| XM_017009436.1 | XP_016864925.1 | 3 | 946 | |||
| XM_017009437.1 | XP_016864926.1 | 3 | 1744 | |||
| XM_017009438.1 | XP_016864927.1 | 3 | 1052 | |||
| GenBank mRNA | AK127142.1 | - | 3 | 997 | ||
| BX648329.1 | - | 3 | 1025 | |||

PLCXD3
Genomic Map