siRNA Details
| Gene Symbol: OR2A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004135.1 | NP_001004135.1 | 1 | 112 | ||
| GenBank mRNA | BC136902.1 | - | 1 | 172 | 9020893 | |
| BC136903.1 | - | 1 | 172 | 9020894 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2A12| Gene Symbol: OR2A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004135.1 | NP_001004135.1 | 1 | 112 | ||
| GenBank mRNA | BC136902.1 | - | 1 | 172 | 9020893 | |
| BC136903.1 | - | 1 | 172 | 9020894 | ||