siRNA Details
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007563.2 | NP_001007564.1 | 4 | 793 | ||
GenBank mRNA | AK131521.1 | - | 4 | 346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IGFBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007563.2 | NP_001007564.1 | 4 | 793 | ||
GenBank mRNA | AK131521.1 | - | 4 | 346 |