siRNA Details
| Gene Symbol: LANCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198511.2 | NP_940913.1 | 5 | 1421 | ||
| GenBank mRNA | AK124915.1 | - | 5 | 1421 | ||
| BC093667.1 | - | 5 | 1213 | 7939512 | ||
| BC093669.1 | - | 5 | 1213 | 7939514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LANCL3| Gene Symbol: LANCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198511.2 | NP_940913.1 | 5 | 1421 | ||
| GenBank mRNA | AK124915.1 | - | 5 | 1421 | ||
| BC093667.1 | - | 5 | 1213 | 7939512 | ||
| BC093669.1 | - | 5 | 1213 | 7939514 | ||