siRNA Details
| Gene Symbol: CATSPER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178019.2 | NP_821138.1 | 7 | 1135 | ||
| GenBank mRNA | AF432876.1 | - | 7 | 1117 | ||
| AK292427.1 | - | 7 | 1134 | |||
| AY156951.1 | - | 3 | 337 | |||
| BC101692.1 | - | 7 | 1095 | 8069198 | ||
| BC110384.1 | - | 7 | 1135 | 5166215 | ||

CATSPER3
Genomic Map