siRNA Details
| Gene Symbol: SLC35B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286509.1 | NP_001273438.1 | 4 | 990 | ||
| NM_001286510.1 | NP_001273439.1 | 4 | 989 | |||
| NM_001286511.1 | NP_001273440.1 | 4 | 944 | |||
| NM_001286512.1 | NP_001273441.1 | 4 | 993 | |||
| NM_001286513.1 | NP_001273442.1 | 3 | 839 | |||
| NM_001286517.1 | NP_001273446.1 | 3 | 800 | |||
| NM_001286519.1 | NP_001273448.1 | 2 | 645 | |||
| NM_001286520.1 | NP_001273449.1 | 4 | 998 | |||
| NM_178148.3 | NP_835361.1 | 4 | 994 | |||
| GenBank mRNA | AB106538.1 | - | 4 | 842 | ||
| AK075456.1 | - | 4 | 923 | |||
| AK123749.1 | - | 4 | 891 | |||
| AK223028.1 | - | 3 | 644 | |||
| AK293251.1 | - | 2 | 612 | |||
| AK293344.1 | - | 3 | 796 | |||
| AK307572.1 | - | 4 | 929 | |||
| AY491520.1 | - | 4 | 960 | |||
| BC016839.1 | - | 1 | 17 | |||
| BC024288.1 | - | 4 | 958 | 4890949 | ||

SLC35B2
Genomic Map