siRNA Details
| Gene Symbol: FGF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002007.2 | NP_001998.1 | 3 | 1099 | ||
| GenBank mRNA | M17446.1 | - | 3 | 1098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF4| Gene Symbol: FGF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002007.2 | NP_001998.1 | 3 | 1099 | ||
| GenBank mRNA | M17446.1 | - | 3 | 1098 | ||