siRNA Details
| Gene Symbol: LINC00442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026852.1 | 3 | 1074 | |||
| GenBank mRNA | AK301801.1 | - | 3 | 1074 | ||
| BC047425.1 | - | 3 | 1074 | |||
| BC067875.1 | - | 3 | 1074 | |||
| BC094789.1 | - | 3 | 1074 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00442| Gene Symbol: LINC00442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026852.1 | 3 | 1074 | |||
| GenBank mRNA | AK301801.1 | - | 3 | 1074 | ||
| BC047425.1 | - | 3 | 1074 | |||
| BC067875.1 | - | 3 | 1074 | |||
| BC094789.1 | - | 3 | 1074 | |||