siRNA Details
Gene Symbol: FGF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002007.2 | NP_001998.1 | 3 | 1045 | ||
GenBank mRNA | AW340506.1 | - | 2 | 377 | ||
M17446.1 | - | 3 | 1044 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FGF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002007.2 | NP_001998.1 | 3 | 1045 | ||
GenBank mRNA | AW340506.1 | - | 2 | 377 | ||
M17446.1 | - | 3 | 1044 |