siRNA Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105069.1 | NP_001098539.1 | 14 | 1862 | ||
| NM_182617.3 | NP_872423.3 | 15 | 1897 | |||
| XM_011545824.2 | XP_011544126.1 | 13 | 1683 | |||
| GenBank mRNA | AB073604.1 | - | 15 | 1828 | ||
| AK129944.1 | - | 14 | 1801 | |||
| AK289416.1 | - | 14 | 1801 | |||
| AY160217.1 | - | 13 | 1687 | |||

ACSM2B
Genomic Map