siRNA Details
| Gene Symbol: SELV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182704.1 | NP_874363.1 | 2, 3 | 941 | ||
| GenBank mRNA | AY324825.1 | - | 2, 3 | 941 | ||
| BC117331.1 | - | 2, 3 | 906 | 40125882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SELV| Gene Symbol: SELV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182704.1 | NP_874363.1 | 2, 3 | 941 | ||
| GenBank mRNA | AY324825.1 | - | 2, 3 | 941 | ||
| BC117331.1 | - | 2, 3 | 906 | 40125882 | ||