siRNA Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042693.2 | NP_001036158.1 | 2 | 458 | ||
| NR_135156.1 | 3 | 551 | ||||
| NR_135157.1 | 2 | 458 | ||||
| XM_011541377.2 | XP_011539679.1 | 2 | 1727 | |||
| XM_011541378.2 | XP_011539680.1 | 2 | 1728 | |||
| GenBank mRNA | AK128236.1 | - | 2 | 458 | ||
| AY358621.1 | - | 2 | 232 | |||
| BC041608.1 | - | 3 | 501 | |||
| BG192125.1 | - | 1 | 130 | |||

FAM159A
Genomic Map