siRNA Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042693.2 | NP_001036158.1 | 2 | 418 | ||
| NR_135156.1 | 3 | 511 | ||||
| NR_135157.1 | 2 | 418 | ||||
| XM_011541377.2 | XP_011539679.1 | 2 | 1687 | |||
| XM_011541378.2 | XP_011539680.1 | 2 | 1688 | |||
| GenBank mRNA | AK128236.1 | - | 2 | 418 | ||
| AY358621.1 | - | 2 | 192 | |||
| BC041608.1 | - | 3 | 461 | |||
| BG192125.1 | - | 1 | 90 | |||

FAM159A
Genomic Map