siRNA Details
| Gene Symbol: GEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130009.2 | NP_001123481.2 | 14 | 1942 | ||
| NM_182625.4 | NP_872431.4 | 14 | 1866 | |||
| XM_005262613.4 | XP_005262670.1 | 14 | 1952 | |||
| XM_006712005.3 | XP_006712068.1 | 14 | 1849 | |||
| XM_011532820.2 | XP_011531122.1 | 14 | 1869 | |||
| XM_011532821.2 | XP_011531123.1 | 14 | 1959 | |||
| XM_011532822.2 | XP_011531124.1 | 14 | 1839 | |||
| GenBank mRNA | AK098188.1 | - | 7 | 892 | ||
| AK131387.1 | - | 14 | 1814 | |||
| BC117204.1 | - | 13 | 1764 | 40125755 | ||
| BC117206.1 | - | 13 | 1764 | 40125757 | ||
| CR936783.1 | - | 14 | 1952 | |||

GEN1
Genomic Map